Learn More
Perkin Elmer US LLC High-Sensitivity GemTip Plastic Body Nebulizer Complete Assembly for AAnalyst
Supplier:  Perkin Elmer US LLC N0370393
Encompass_Preferred
A complete GemTip High-Sensitivity plastic body nebulizer featuring Kalrez o-rings and plastic needle with Sapphire tip. This nebulizer was designed to obtain optimum sensitivity, detection limits and precision with flame atomic absorption for both air-acetylene and nitrous oxide-acetylene flames. This assembly includes: Capillary Tubing, Qty. 1 (09908265) Fine Adjustment Nut, Qty. 1 (N0371492) Gland Nut, Qty. 1 (N0371505) PTFE Tubing, Qty. 1 (09985708) Capillary Assembly for High-Sensitivity GemTip Nebulizers, Qty. 1 (N0371522) O-Ring for Capillary Assembly, Qty. 1 (09926083) Impact Bead for High-Sensitivity GemTip Nebulizer, Qty. 1 (B0505086) O-Ring for Nebulizer Body, Qty. 1 (09902015) Body Assembly for High-Sensitivity Nebulizer, Qty. 1 (N0370399) Locking Nut, Qty. 1 (N0371493) Spring, Qty. 1 (09902473) Connector Nut, Qty. 1 (N0371485) Magnet Holder Assembly, Qty.1 (N1010215) This nebulizer is compatible with the PerkinElmer AAnalyst 200/400*, AAnalyst100/300/700/800, and Models
Catalog No. 50-904-3583