Learn More
Abcam High-Sensitivity ChIP Kit, 24 Tests
Catalog No. 502590282
Encompass_Preferred
High-sensitivity ChIP kit is a complete set of optimized reagents to carry out a successful chromatin immunoprecipitation procedure in a high throughput format starting from mammalian cells or tissues. - Cited in over 40 publications - Optimized to reduce non-specific background - Enables detection of low-abundance targets - Compatible with PCR, microarrays & sequencing
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-259-0282 Supplier Abcam Supplier No. AB18591324TEST
High-sensitivity ChIP kit is a complete set of optimized reagents to carry out a successful chromatin immunoprecipitation procedure in a high throughput format starting from mammalian cells or tissues. - Cited in over 40 publications - Optimized to reduce non-specific background - Enables detection of low-abundance targets - Compatible with PCR, microarrays & sequencing
The product is subject to the following: Abcam Restricted Use Statement