Learn More
Agilent Technologies High Sensitivity D1000 Reagents
Catalog No. NC1860068
Encompass
For the high sensitivity analysis of DNA from 35 to 1000 bp. Includes ladder and sample buffer.
Catalog No. NC1860068 Supplier Agilent Technologies Supplier No. 50675585
For the high sensitivity analysis of DNA from 35 to 1000 bp. Includes ladder and sample buffer.