Learn More
Agilent Technologies High Sensitivity D1000 Sample Buffer
Catalog No. NC1874889
Encompass
For the high sensitivity analysis of DNA from 35 to 1000 bp. Sample Buffer only.
Catalog No. NC1874889 Supplier Agilent Technologies Supplier No. 50675603
For the high sensitivity analysis of DNA from 35 to 1000 bp. Sample Buffer only.