Learn More
Waters Corp Waters High Sensitivity Filter Mixer, 425μL, 10,000 psi, for Peptides
Supplier:  Waters Corp 205000403
Encompass_Preferred
The Waters High Sensitivity Filter Mixer is specifically designed for ACQUITY UPLC Binary Solvent Managers. It handles long, shallow gradients that require increased mixing, and is particularly suited for peptide analysis, with a pressure rating of 10,000 psi.
  • Designed for long, shallow gradients that necessitate increased mixing.
  • Rated for high-pressure applications up to 10,000 psi.
  • Optimized for use with ACQUITY UPLC Binary Solvent Managers.
  • Ideal for peptide analysis.
  • Features a 425μL volume for precise mixing.
Catalog No. 50-203-9225