Learn More
Perkin Elmer US LLC High Sensitivity Nebulizer End Cap Assembly with Kalrez O-Rings for PinAAcle
Catalog No. 502104339
Encompass_Preferred
This end cap assembly is included in the Organics High Sensitivity Plastic Nebulizer Complete Assembly with Pt/Ir Needle for PinAAcle (N3160159) and Organics High Sensitivity Plastic Nebulizer Complete Assembly with Tantalum Needle for PinAAcle (N3160152). Note: This end cap ships as standard equipment on instruments configured with a high sensitivity nebulizer.
Catalog No. 50-210-4339 Supplier Perkin Elmer US LLC Supplier No. N3160189
This end cap assembly is included in the Organics High Sensitivity Plastic Nebulizer Complete Assembly with Pt/Ir Needle for PinAAcle (N3160159) and Organics High Sensitivity Plastic Nebulizer Complete Assembly with Tantalum Needle for PinAAcle (N3160152). Note: This end cap ships as standard equipment on instruments configured with a high sensitivity nebulizer.