Learn More
Perkin Elmer US LLC High Sensitivity Plastic Nebulizer Assembly with Chemical Resistant O-Rings for PinAAcle 900
Catalog No. 502112808
Encompass_Preferred
The high sensitivity plastic nebulizer is an adjustable pneumatic device that is used to draw the sample into the needle assembly tubing and spray it as a fine aerosol into the spray chamber.
Catalog No. 50-211-2808 Supplier Perkin Elmer US LLC Supplier No. N3160187
The high sensitivity plastic nebulizer is an adjustable pneumatic device that is used to draw the sample into the needle assembly tubing and spray it as a fine aerosol into the spray chamber.