Learn More
Agilent Technologies High Sensitivity Protein 250 Kit
Catalog No. NC1846969
Encompass
For sizing and highly sensitive quantitation of protein samples from 10 to 250 kDa. The kit includes 10 microfluidic chips, reagents and consumables, sufficient for 100 samples.
Catalog No. NC1846969 Supplier Agilent Technologies Supplier No. 50671575
For sizing and highly sensitive quantitation of protein samples from 10 to 250 kDa. The kit includes 10 microfluidic chips, reagents and consumables, sufficient for 100 samples.