Learn More
Perkin Elmer US LLC High Sensitivity Quartz Cyclonic Spray Chamber with Matrix Gas Port for NexION 1000/2000
Catalog No. 502112156
Encompass_Preferred
The spray chamber is an integral part of the sample introduction system responsible for filtering the sample mist to permit the appropriate droplet size distribution to reach the plasma. The baffled cyclonic spray chamber features a central transfer tube which acts as a secondary particle separator helping separate larger aerosol particles from the sample. This reduces solvent load in the plasma without compromising detection limits. The distance between the bottom of the baffle and the chamber wall is carefully controlled so that droplets cannot form on the bottom of the baffle. The advantage is smooth draining and improved precision. This spray chamber is included in the Organics Sample Introduction Kit (Orange) for NexION 1000/2000 (N8150031).
Catalog No. 50-211-2156 Supplier Perkin Elmer US LLC Supplier No. N8152383
The spray chamber is an integral part of the sample introduction system responsible for filtering the sample mist to permit the appropriate droplet size distribution to reach the plasma. The baffled cyclonic spray chamber features a central transfer tube which acts as a secondary particle separator helping separate larger aerosol particles from the sample. This reduces solvent load in the plasma without compromising detection limits. The distance between the bottom of the baffle and the chamber wall is carefully controlled so that droplets cannot form on the bottom of the baffle. The advantage is smooth draining and improved precision. This spray chamber is included in the Organics Sample Introduction Kit (Orange) for NexION 1000/2000 (N8150031).