Learn More
Agilent Technologies High Sensitivity RNA ScreenTape Sample Buffer.
Catalog No. NC1869822
Encompass
High Sensitivity RNA ScreenTape Sample Buffer. For the high sensitivity analysis of total RNA down to a sensitivity of 100 pg/L. Sample Buffer only.
Catalog No. NC1869822 Supplier Agilent Technologies Supplier No. 50675580
High Sensitivity RNA ScreenTape Sample Buffer. For the high sensitivity analysis of total RNA down to a sensitivity of 100 pg/L. Sample Buffer only.