Learn More
Agilent Technologies High Sensitivity RNA ScreenTape Ladder, Ladder Only
Catalog No. NC1869828
Encompass
High Sensitivity RNA ScreenTape Ladder. For the high sensitivity analysis of total RNA down to a sensitivity of 100 pg/L.
Catalog No. NC1869828 Supplier Agilent Technologies Supplier No. 50675581
High Sensitivity RNA ScreenTape Ladder. For the high sensitivity analysis of total RNA down to a sensitivity of 100 pg/L.