Learn More
BUCHI High Shear Disperser HSD
SDP
Catalog No. 05376250 Shop All BUCHI Products
Change view
Click to view available options
Product Type:
Digital Disperser
Catalog No. Product Type
05-376-250 Digital Disperser
1 options
Catalog No. 05-376-250 Supplier BUCHI Supplier No. 11HSD300N02
Add to Cart
Add to Cart
High Shear Disperser HSD for precise, reliable, flexible sample preparation in demanding lab workflows.
  • Powerful Motor—High-efficiency motor ensures consistent dispersing performance across a wide range of viscosities.
  • Wide Speed Range—Digital regulation from 3,000 to 25,000 rpm, suitable for everything from gentle mixing to intense homogenization.
  • Digital Display—Clear screen provides real-time speed monitoring and easy adjustments for reproducible results.
  • Versatile Tool Compatibility—Compatible with a broad selection of dispersing shafts and rotors for various sample types and volumes.
  • Volume Range—Capable of processing samples from 0.25 mL up to 30,000 mL (30 L) depending on the chosen accessory.
  • Robust and Durable—Designed with high-quality, chemically resistant materials for long-lasting laboratory use.
  • Safety Features—Overload protection and stable construction ensure safe operation during high-speed runs.
Benefits
  • Provides accurate, reproducible homogenization for a wide variety of applications.
  • Ensures operator safety with built-in protection systems and stable design.
  • Saves time and effort with digital speed control and intuitive operation.
  • Highly versatile—processes both very small and very large sample volumes.
  • Long-lasting and maintenance-free motor reduces downtime in busy labs.
Specifications
Voltage 115 VAC
Product Type Digital Disperser