Learn More
MSC Spartan Chemical iShine Floor Finish, 25% High Solids, 5-gallon Pail
Supplier: MSC 57184525
Spartan Chemical iShine Floor Finish is a high-solids formula designed to provide exceptional gloss, clarity, and durability for floors. Featuring 25% non-volatile solids and patented optically enhanced polymer technology, this finish offers maximum protection against scuff marks, water spots, and powdering while ensuring a non-yellowing, crystal-clear appearance. Its quick-building formula allows for superior protection in just four coats, saving on labor costs and ensuring ease of application with incredible leveling.
- Amplifies overall gloss and clarity
- Exceptional wear and gloss retention
- Non-yellowing, crystal-clear finish
- Resists scuffmarks, water spots, and black heel marks
- Quick-building formula saves labor time
- Easy application with incredible leveling
- UL Approved for slip resistance
- Withstands three freeze/thaw cycles