Learn More
MTC Bio High Speed Centrifuge Tube 10mL Oak Ridge Style Round Bottom 50 000xg Max RCF w/ Screw Cap Resealable Bag 12/pk
Catalog No. 5000575210
Encompass_Preferred
High Speed Centrifuge Tube 10mL Oak Ridge Style Round Bottom 50 000xg Max RCF w/ Screw Cap Resealable Bag 12/pk
Catalog No. 50-005-75210 Supplier MTC Bio Supplier No. C0100
High Speed Centrifuge Tube 10mL Oak Ridge Style Round Bottom 50 000xg Max RCF w/ Screw Cap Resealable Bag 12/pk