Learn More
MTC Bio High Speed Centrifuge Tube 15mL Conical Bottom 50 000xg Max RCF Graduated w/ Screw Cap and Gripping Tab Resealable Bag 12/pk
Catalog No. 5000575206
Encompass_Preferred
High Speed Centrifuge Tube 15mL Conical Bottom 50 000xg Max RCF Graduated w/ Screw Cap and Gripping Tab Resealable Bag 12/pk
Catalog No. 50-005-75206 Supplier MTC Bio Supplier No. C0300
High Speed Centrifuge Tube 15mL Conical Bottom 50 000xg Max RCF Graduated w/ Screw Cap and Gripping Tab Resealable Bag 12/pk