Learn More
MTC Bio High Speed Centrifuge Tube 30mL Oak Ridge Style Round Bottom 50 000xg Max RCF w/ Screw Cap and Gripping Tab Resealable Bag 12/pk
Catalog No. 5000575212
Encompass_Preferred
High Speed Centrifuge Tube 30mL Oak Ridge Style Round Bottom 50 000xg Max RCF w/ Screw Cap and Gripping Tab Resealable Bag 12/pk
Catalog No. 50-005-75212 Supplier MTC Bio Supplier No. C0108
High Speed Centrifuge Tube 30mL Oak Ridge Style Round Bottom 50 000xg Max RCF w/ Screw Cap and Gripping Tab Resealable Bag 12/pk