Learn More
MTC Bio High Speed Centrifuge Tube 35mL Conical Bottom 50 000xg Max RCF Graduated w/ Screw Cap and Gripping Tab Resealable Bag 12/pk
Catalog No. 5000575214
Encompass_Preferred
High Speed Centrifuge Tube 35mL Conical Bottom 50 000xg Max RCF Graduated w/ Screw Cap and Gripping Tab Resealable Bag 12/pk
Catalog No. 50-005-75214 Supplier MTC Bio Supplier No. C0302
High Speed Centrifuge Tube 35mL Conical Bottom 50 000xg Max RCF Graduated w/ Screw Cap and Gripping Tab Resealable Bag 12/pk