Learn More
MTC Bio High Speed Centrifuge Tube 50mL Oak Ridge Style Round Bottom 50 000xg Max RCF w/ Screw Cap and Gripping Tab Resealable Bag 12/pk
Catalog No. 5000575213
Encompass_Preferred
High Speed Centrifuge Tube 50mL Oak Ridge Style Round Bottom 50 000xg Max RCF w/ Screw Cap and Gripping Tab Resealable Bag 12/pk
Catalog No. 50-005-75213 Supplier MTC Bio Supplier No. C0110
High Speed Centrifuge Tube 50mL Oak Ridge Style Round Bottom 50 000xg Max RCF w/ Screw Cap and Gripping Tab Resealable Bag 12/pk