Learn More
RWD LIFE SCIENCE INC High-Speed Benchtop Refrigerated Centrifuge M1416R, Max Speed 18,000 rpm (31,152×g), Capacity 24 x 2mL/4 x 400mL
SDP
Catalog No. NC3404386
Encompass_Preferred
M1416R centrifuge is a refrigerated model with the speed up to 18,000 rpm (31,152×g), adapting various applications from micro to medium throughput. The capacity of M1416R ranges from 24 x 2mL to 4 x 400mL, and the plate rotor also accommodates PCR and deep-well plates. In addition, equipped with a 7-inch touch screen, M1416R provides visual operation and a personalized account management system ensuring efficient and quality control.
Catalog No. NC3404386 Supplier RWD LIFE SCIENCE INC Supplier No. M1416R
Add to Cart
Add to Cart
Request a Quote
M1416R centrifuge is a refrigerated model with the speed up to 18,000 rpm (31,152×g), adapting various applications from micro to medium throughput. The capacity of M1416R ranges from 24 x 2mL to 4 x 400mL, and the plate rotor also accommodates PCR and deep-well plates. In addition, equipped with a 7-inch touch screen, M1416R provides visual operation and a personalized account management system ensuring efficient and quality control.