Learn More
Genscript Corporation High-Stability PCR Kit
Catalog No. 5000058365
Encompass
GenScript High-Stability PCR Kit contains Green i Taq /i DNA polymerase 10 Taq buffer and Stabilized dNTP Mix 10 mM Each all specially formulated for increased stability using our proprietary technology The kit can be stored at 4 for over six months or at room temperature (25 ) for one month without significant loss of activity The stabilizers in the buffers also increase the stability of Green Taq DNA polymerase at higher temperature (72 ) increasing the DNA yield of PCR amplifications of long DNA This kit facilitates quick and easy PCR setup without any need for thawing This kit can also be used in high-throughput assays in which all the PCR reagents need to be ready and stable for PCR setup at any time
Catalog No. 50-000-58365 Supplier Genscript Corporation Supplier No. L00342
GenScript High-Stability PCR Kit contains Green i Taq /i DNA polymerase 10 Taq buffer and Stabilized dNTP Mix 10 mM Each all specially formulated for increased stability using our proprietary technology The kit can be stored at 4 for over six months or at room temperature (25 ) for one month without significant loss of activity The stabilizers in the buffers also increase the stability of Green Taq DNA polymerase at higher temperature (72 ) increasing the DNA yield of PCR amplifications of long DNA This kit facilitates quick and easy PCR setup without any need for thawing This kit can also be used in high-throughput assays in which all the PCR reagents need to be ready and stable for PCR setup at any time