Learn More
SPECTRAPURE INC HIGH TEMP MIXED BED DI CART
Catalog No. NC1978136
Encompass
High Temp Mixed Bed DI Cart 2.5in dia x 6in L w/ 100um Filter Pads
Catalog No. NC1978136 Supplier SPECTRAPURE INC Supplier No. DIMBHTQ2.5X6L21
High Temp Mixed Bed DI Cart 2.5in dia x 6in L w/ 100um Filter Pads