Learn More
Formlabs Inc High Temp Resin, 1L
SDP
Catalog No. NC1692628
Encompass
High Temp Resin offers a heat deflection temperature (HDT) of 238 °C @ 0.45 MPa, the highest among Formlabs resins. Use it to print detailed, precise prototypes with high temperature resistance.
Catalog No. NC1692628 Supplier Formlabs Inc Supplier No. RSF2HTAM02
High Temp Resin offers a heat deflection temperature (HDT) of 238 °C @ 0.45 MPa, the highest among Formlabs resins. Use it to print detailed, precise prototypes with high temperature resistance.