Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ High Temperature Aluminum Foil/Glass Cloth Tape

Catalog No. 19070904 Shop All 3M Products
Change view
Click to view available options
Material:
Aluminum Foil, Glass Cloth
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-070-904 Aluminum Foil, Glass Cloth
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-070-904 Supplier 3M™ Supplier No. 7000028867
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

3M Company High Temperature Aluminum Foil/Glass Cloth Tape offers puncture, fracture and tear resistance

  • Performs continuously over a wide temperature range from -65°F (-54°C) to 600°F (316°C)
  • Light and flexible construction easily conforms to curved or irregular surfaces
  • Thermally conductive and reflective backing assists with heat shielding
  • Pressure sensitive silicone adhesive adheres to difficult surfaces, including silicone
  • Consists of an aluminum foil, glass cloth backing combined with a pressure sensitive silicone adhesive
  • Ideal for use as a protective wrap for heat sensitive cables and other instruments exposed to movement and vibration
  • Commonly used in aerospace, automotive and general industrial applications
  • 2 in x 36 yd 7.3 mil
  • 3M Catalog# 363

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Shielding cables and wire harnesses against heat
Color Silver
Material Aluminum Foil, Glass Cloth
Model 363
Quantity 24 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.