Learn More
Stoelting™ High Temperature Disposable Cautery Pens
SDP
Catalog No. 10001020 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Fine tip
Catalog No. Model
10-001-020 Fine tip
1 options
Catalog No. 10-001-020 Supplier Stoelting™ Supplier No. 59016
Add to Cart
Add to Cart
Stoelting™ High Temperature Disposable Cautery Pens are ideal for procedures including, pinpoint hemostasis, controlling diffuse bleeders, vasectomies, and sculpting woven grafts.
Individually packaged sterile, 10 per box.
Specifications
For Use With (Application) Neuroscience
Temperature (Metric) Range Up to 1204°C
Model Fine tip
Includes 10/box