Learn More
Alkali Scientific High Temperature Red Caps, up to 180 Degrees Celsius, Thread 45, 10/cs
SDP
Catalog No. 502388982
Encompass_Preferred
High Temperature Red Caps for Hybexª Media Storage Bottles can stand temperatures up to 180℃. Thread 45, ten per pack.
Catalog No. 50-238-8982 Supplier Alkali Scientific Supplier No. RS70209HTC
High Temperature Red Caps for Hybexª Media Storage Bottles can stand temperatures up to 180℃. Thread 45, ten per pack.