Learn More
Hoefer, Inc Hoefer™ SUBHT High Throughput Submarine Gel Electrophoresis Unit
SDP
Catalog No. NC2741111
Encompass_Preferred
Ideal for high throughput screening applications like single strand polymorphism (SSP) analysis following PCRFeatures & Benefits:Fast, error-free loading—Load 96-well plate using a multichannel pipetteSimplified analysis—Fluorescent labels on a UV transparent gel tray, allows for instant sample identification during post-run visualizationHigh throughput—Run up to 120 samples in 30 minutesBetter reproducibility—Eliminate variability caused by running buffer pH drift and ionic gradient formation with optional buffer recirculation capabilityMaximum flexibility—Wide range of combs and accessories to meet analysis needsIncludes:Buffer Chamber with Lid; High Voltage Leads; Fluorescent Labeled Running Tray with Casting Gates; 30 well combs, 4 pcs; Buffer Recirculation PortsSpecifications:Gel Dimensions (W x L): 15 x 15 cmMaximum Buffer Volume: 1.2 LMaximum Power Settings: 300 V, 200 mA, 60 WMaximum Sample Capacity: 120Unit Dimensions (W x L x H): 21.5 x 33.5 x 7 cm
Catalog No. NC2741111 Supplier Hoefer, Inc Supplier No. SUBHT
Ideal for high throughput screening applications like single strand polymorphism (SSP) analysis following PCRFeatures & Benefits:Fast, error-free loading—Load 96-well plate using a multichannel pipetteSimplified analysis—Fluorescent labels on a UV transparent gel tray, allows for instant sample identification during post-run visualizationHigh throughput—Run up to 120 samples in 30 minutesBetter reproducibility—Eliminate variability caused by running buffer pH drift and ionic gradient formation with optional buffer recirculation capabilityMaximum flexibility—Wide range of combs and accessories to meet analysis needsIncludes:Buffer Chamber with Lid; High Voltage Leads; Fluorescent Labeled Running Tray with Casting Gates; 30 well combs, 4 pcs; Buffer Recirculation PortsSpecifications:Gel Dimensions (W x L): 15 x 15 cmMaximum Buffer Volume: 1.2 LMaximum Power Settings: 300 V, 200 mA, 60 WMaximum Sample Capacity: 120Unit Dimensions (W x L x H): 21.5 x 33.5 x 7 cm