Learn More
GeneCopoeia HIGH TITER PURIFIED AAV PRTCL
SDP
Catalog No. NC3593608
Encompass
AAPeB-EGFP-AV13-200-h - quote #20250227-193810471high titer purified AAV particles for eGFP in pEZ-AV13 with hSYN promoter; AAVPHP.eB serotype; 2mL (100uL x 2 vials, >10^13 GC/mL)
Catalog No. NC3593608 Supplier GeneCopoeia Supplier No. AAPEBEGFPAV13200H
AAPeB-EGFP-AV13-200-h - quote #20250227-193810471high titer purified AAV particles for eGFP in pEZ-AV13 with hSYN promoter; AAVPHP.eB serotype; 2mL (100uL x 2 vials, >10^13 GC/mL)