Learn More
GeneCopoeia HIGH TITER PURIFIED AAV PRTCLS
SDP
Catalog No. NC3590343
Encompass
AAPeB-A0813-AV30-s20000-B00-h/Q#20250225-001914276high titer purified AAV particles for human IFNA2 (NM_000605.4) with GFAP pro‐moter; AAV-PHP.eB serotype; 2mL (100uL x 20 vials, >10^13 GC/mL)
Catalog No. NC3590343 Supplier GeneCopoeia Supplier No. AAPEBA0813AV30S20000B
AAPeB-A0813-AV30-s20000-B00-h/Q#20250225-001914276high titer purified AAV particles for human IFNA2 (NM_000605.4) with GFAP pro‐moter; AAV-PHP.eB serotype; 2mL (100uL x 20 vials, >10^13 GC/mL)