Learn More
Supertek High UV C linear lamp
Catalog No. S001285 Shop All Supertek Scientific Products
Change view
Click to view available options
Wattage:
8 W
25 W
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wattage
S001285 8 W
S001284 25 W
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S001285 Supplier Supertek Supplier No. CH12040S8W
Add to Cart
Add to Cart
Designed for effective and environment friendly disinfection without chemicals.
  • Compact efficient germicidal light source designed for effective ultraviolet disinfection applications
  • Emits shortwave UVC radiation at approximately 254 nm to inactivate bacteria viruses and other harmful microorganisms on exposed surfaces
  • 8W UVC output for effective germicidal performance
  • Linear tube design for broad exposure coverage
  • Emits UVC wavelength 254 nm for disinfection applications
  • Suitable for enclosed sterilization systems
  • Energy efficient and long service life
Specifications
Base Style 2-pin
Gas Fill Neon-mercury
Light Source UV-C
Voltage 110/220 V
Wattage 8 W
Color White