Learn More
Fischer Technical Company High Vacuum Hand Held Digital Gauge
SCI_ED
Catalog No. S2337151 Shop All Fischer Technical Company Products
Change view
Click to view available options
Measuring Range:
100,000 to 10 μm
Catalog No. Measuring Range
S2337151 100,000 to 10 μm
1 options
Prop 65
Catalog No. S2337151 Supplier Fischer Technical Company Supplier No. HVG001
Add to Cart
Add to Cart
Prop 65
Digital Vacuum Gauge. Vacuum reading range 100,000 to 10 microns. Measures in Microns, PSIA, inHg, Pascals, Torr, mTorr, and mBar.
  • The improved HVG-001 Digital Vacuum Gauge is an easy-to-use, compact vacuum gauge featuring a sleek new design with enhanced user settings and an extended range sensor.
  • Features an extended range MEMS sensor
  • Vacuum reading range 100,000 to 10 microns
  • Easy-to-read backlit display
  • Improved response time for instantaneous vacuum reading
  • Allows for direct connection to the A/C unit without a hose
  • Audible and visual alarms when target vacuum level is achieved
  • Intuitive user menu with custom setting options
  • Low battery indicator with automatic shutoff
  • Measures in Microns, PSIA, inHg, Pascals, Torr, mTorr and mBar
Certifications
SciEd
Specifications
Product Type Digital Vacuum Gauge
Measuring Range 100,000 to 10 μm
Applications Measuring Vacuum
Description Direct connection to the A/C unit without a hose
Includes Extended range MEMS sensor
Pressure Range 10 to 100,000 μ
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov