Learn More
Synthware™ High Vacuum Metering Plug and Control Knob
GREENER_CHOICESDP
Catalog No. 31502337 Shop All Kemtech Products
Change view
Click to view available options
Size:
0 to 12 mm
0 to 4 mm
0 to 8 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
31-502-337 0 to 4 mm
31-502-338 0 to 8 mm
31-502-339 0 to 12 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-502-337 Supplier Synthware™ Supplier No. S660004T
Add to Cart
Add to Cart
PTFE-protected metering plug and control knob features teflon covered O-rings for increased durability.
  • PTFE protected high vacuum metering plug and control knob
  • Teflon-covered O-rings offer increased protection
  • Fully enclosed
Specifications
Description Fully Enclosed
For Use With (Application) Vacuum Sealing
Includes Teflon Coated O-Rings
Material PTFE
Size 0 to 4 mm