Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fischer Technical Company High Vacuum Pump Oil
SCI_ED

Catalog No. p-7235502 Shop All Fischer Technical Company Products
Change view
Click to view available options
Quantity:
12 Quarts/Cs.
2 Quarts/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S2337139 12 Quarts/Cs.
S2337140 2 Quarts/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S2337139 Supplier Fischer Technical Company Supplier No. OIL012
Only null left
Add to Cart
Add to Cart

High Vacuum Pump Oil for use in any Fischer Technical LAV-Series Vacuum Pump.

  • High Vacuum Pump Oil
  • For use in any Fischer Technical LAV-Series vacuum pumps
  • Our oil is extremely pure and non-detergent. It is hydro processed, meaning it goes through a series of catalytic steps rendering the oil extremely refined, more viscous, and more stable.
  • The result is clear mineral oil which will alert you to contamination sooner as it becomes cloudy or milky.
  • In extreme temperatures, the oil will be more resistant to contaminants.

Certifications

SciEd

Specifications

Product Type Vacuum Pump Oil
Format Oil
For Use With (Application) High Vacuum
For Use With (Equipment) Any Fischer Technical LAV-Series Vacuum Pumps
Quantity 12 Quarts/Cs.
Volume (English) 12 qt.
Volume (Metric) 11,356.2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.