Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fischer Technical Company™ Acrylic High Vacuum Pump Plate
SCI_ED

Catalog No. s64988 Shop All Fischer Technical Company Products
Change view
Click to view available options
Type:
Vacuum Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
S64988 Vacuum Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S64988 Supplier Fischer Technical Company™ Supplier No. AP001A
Only null left
Add to Cart
Add to Cart

This affordable quality high vacuum pump plate can accommodate bell jars up to 9 inches in diameter.

  • Made of acrylic and does not require reinforcing ribbons, which limit the size of bell jars you can use
  • The 0.38 in. hose barb connection is compatible with most vacuum pumps
  • The 10.75 x 1.5 in. base has four suction cups to hold the plate in place
  • Replacement gaskets available

Specifications

For Use With 9 in. dia. Bell Jar
Type Vacuum Plate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.