Learn More
Fischer Technical Company High Vacuum Pump Repair Parts
SCI_ED
Catalog No. S2337147 Shop All Fischer Technical Company Products
Change view
Click to view available options
Flow Rate:
None
Catalog No. Flow Rate
S2337147 None
1 options
Catalog No. S2337147 Supplier Fischer Technical Company Supplier No. LAVREPAIRKIT
Add to Cart
Add to Cart
Vacuum Pump Repair Kit includes Shaft Seal, Cover Seal, Flexible Coupler, Cartridge Valve, and High Vacuum Pump Oil.
  • This repair kit includes all the essential items which may require replacement when performing a general maintenance on the LAV series High Vacuum Pumps.
  • The kit includes 1 ea. PR-3 (Shaft Seal), 1 ea. PR-311 (Cover Seal), 1 ea. PR-208 (Flexible Coupler), 1 ea. PR-18 (Cartridge Valve), 2 ea. OIL-001 (Quart of Oil)
Certifications
SciEd
Specifications
Includes Shaft Seal, Cover Seal, Flexible Coupler, Cartridge Valve, and High Vacuum Pump Oil
Flow Rate None
Pump Type Two-stage Rotary Vane Vacuum Pump
Applications LAV Series High Vacuum Pump Repair
Maximum Pressure None
Voltage 0/1 V
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov