Learn More
Fischer Technical Company High Vacuum Pump Repair Parts
SCI_ED
Catalog No. S2337141 Shop All Fischer Technical Company Products
Change view
Click to view available options
Flow Rate:
None
Catalog No. Flow Rate
S2337141 None
1 options
Prop 65
Catalog No. S2337141 Supplier Fischer Technical Company Supplier No. PR208
Add to Cart
Add to Cart
Prop 65
Flexible Coupler for LAV Series Pumps
  • Replacement Flexible Coupler for Fischer Technical LAV Series Pumps.
  • The cylindrical coupler kit comes with end cap and set screws to connect the motor to the cartridge shaft of the vacuum pump.
  • Besides allowing the motor to turn the cartridge rotors it is designed to fail and break in the event that some solid gets inside the pump cartridge and locks up the rotors in order to avoid any further internal damage to the pump.
Certifications
SciEd
Specifications
Flow Rate None
Pump Type High Vacuum Pump
Applications LAV Serieds High Vacuum Repair
Maximum Pressure None
Voltage 0/1 V