Learn More
Fischer Technical Company High Vacuum Shut Off and Vacuum Release Valve
SCI_ED
Catalog No. S2337150 Shop All Fischer Technical Company Products
Change view
Click to view available options
Flow Rate:
None
Catalog No. Flow Rate
S2337150 None
1 options
Prop 65
Catalog No. S2337150 Supplier Fischer Technical Company Supplier No. LAV3WAYVALVE
Add to Cart
Add to Cart
Prop 65
High Vacuum Shut off and Vacuum Release Valve
  • Our three-way brass ball valve with T-full port and 3 ea. 3/8 in. Hosebarb connections will hold a vacuum in your system, isolate the Vacuum Pump to prevent oil backup, and release/bleed air back into your system all in one.
  • An external Vacuum Gauge can be connected to the third valve to take a system vacuum reading isolated from the Vacuum Pump.
Certifications
SciEd
Specifications
Description Three-way brass ball valve with T-full port
Flow Rate None
Pump Type High Vacuum Pump
Applications High Vacuum
Maximum Pressure None
Voltage 0/1 V