Learn More
Synthware™ High Vacuum Valve Plug, Viton™ O-ring
GREENER_CHOICESDP
Catalog No. 31502055 Shop All Kemtech Products
Change view
Click to view available options
Size:
0 to 12 mm
0 to 4 mm
0 to 8 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
31-502-055 0 to 12 mm
31-502-053 0 to 4 mm
31-502-054 0 to 8 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-502-055 Supplier Synthware™ Supplier No. S540012
Add to Cart
Add to Cart
Complete with control knob and PTFE stem having three O-rings.
Specifications
Actuation Manual
For Use With (Application) Organic Synthesis
Material Borosilicate Glass
Type High Vacuum Plug and Control Knob
Size 0 to 12 mm