Learn More
Synthware™ High Vacuum Valve Plug, PTFE Protected
GREENER_CHOICESDP
Catalog No. 31502067 Shop All Kemtech Products
Change view
Click to view available options
Size:
0 to 12 mm
0 to 8 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
31-502-067 0 to 12 mm
31-502-066 0 to 8 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31-502-067 Supplier Synthware™ Supplier No. S600012
Add to Cart
Add to Cart
Complete with control knob. Viton™ O-rings on PTEF shaft are protected with precision PTFE wipers.
  • This design provides for an excellent vacuum tight seal without exposing any material other than PTFE to chemicals.
Specifications
Actuation Manual
For Use With (Application) Organic Synthesis
Material Borosilicate Glass
Type High Vacuum Valve
Size 0 to 12 mm