Learn More
TA Instruments High Viscosity Standard Oil S600, 1100 mPa.s (cP) at 25 Degrees Celsius
Catalog No. 5000580627 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments High Viscosity Standard Oil S600 has a viscosity of approximately 1100 mPa.s (cP) at 25 degrees Celsius.
Catalog No. 50-005-80627 Supplier TA Instruments Supplier No. 250024.002
Waters TA Instruments High Viscosity Standard Oil S600 has a viscosity of approximately 1100 mPa.s (cP) at 25 degrees Celsius.