Learn More
Lakeland Industries High Visibility
Supplier:  Lakeland Industries AJPVC10LY3X
Encompass_Preferred
Lakeland Arc X PVC Rain Jackets are comfortable and durable rainwear that offers protection against electric arc flash and the harsh elements. AJPVC10LY jackets are configured with an oversized hood, D-ring pass-through, high-frequency sealed seams, and vented back. Available in sizes SM-5X, this lighter weight jacket is ideal for working in cold, rainy weather when electric and flash fires are of concern.
Catalog No. 50-284-0920