Learn More
Lakeland Industries High Visibility
Catalog No. 502841049
Encompass_Preferred
Lakeland's flame resistant, hi-visibility coveralls are made with Westex DH 6.5 oz fabric for active moisture wicking capabilities and a soft, comfortable feel. Paired with 3M Scotchlite Reflective Trim, this high visibility FR coverall is a great choice when a job calls for visibility compliance.
Catalog No. 50-284-1049 Supplier Lakeland Industries Supplier No. TSP010LSM30
Lakeland's flame resistant, hi-visibility coveralls are made with Westex DH 6.5 oz fabric for active moisture wicking capabilities and a soft, comfortable feel. Paired with 3M Scotchlite Reflective Trim, this high visibility FR coverall is a great choice when a job calls for visibility compliance.