Learn More
Lakeland Industries High Visibility
Supplier:  Lakeland Industries VAMC2GDKWLXL
Encompass_Preferred
Made with polyester mesh and trimmed with 2 inch silver material, this Class 2 hi-vis vest is affordable, comfortable, and safe. This vest features an inside pocket along with an outer pencil pocket and zipper closure. *Please note that this vest is not FR treated.
Catalog No. 50-284-1196