Learn More
Lakeland Industries High Visibility
Supplier:  Lakeland Industries ABPVC10LY4X
Encompass_Preferred
Lakeland Arc X PVC Bib Style Rain Pants are comfortable and durable rainwear that offers protection against electric arc flash and the harsh elements. Available in sizes SM-4X, this protective bib pant is ideal for working in cold, rainy weather when electric and flash fires are of concern.
Catalog No. 50-284-0852