Learn More
Lakeland Industries High Visibility
Supplier:  Lakeland Industries V8AM0112VL3X
Encompass_Preferred
Lakeland Class 2 FR Modacrylic Mesh Hi-Vis Vests are designed with FR treated mesh fabric, and reflective material. Available in sizes SM-5X, these vests are ideal for worker environments where visibility is important, and when flash fire is a concern.
Catalog No. 50-284-1135