Learn More
Lakeland Industries High Visibility
Supplier:  Lakeland Industries ABPU10LYZ3X
Encompass_Preferred
More Comfortable Cooler: Breathes and releases perspiration. Soft and more flexible Lighter weight More Durable for Longer Wear Cycle Softness and pliability make it less prone to get caught on branches and brush Superior puncture, tear and abrasion resistance Reinforced HF welded, sealed and taped seams for maximum strength Innovative DRP Diamond Reinforced Patch to prevent crotch ?blow out" Saves money in the long run Certification: Jacket: ANSI 107-2010 Class 3 Bib Overall: ANSI 107-2010 Class E ASTM F1891 ASTM F903 Fabric information: 10.0 oz.² Polyurethane over FR cotton knit Arc Rating: 16.3 Cal/cm² Ebtas: 21.9 Cal/cm?² HAF: 84% MVTR: 305G/M?²/24H
Catalog No. 50-284-1164