Learn More
Lakeland Industries High Visibility
Supplier:  Lakeland Industries VAFOSP2GBVLR
Encompass_Preferred
Class, 2 5-Point Breakaway Public Safety Premium Vest - Solid 5-Point Breakaway Public Safety Premium Vest is designed with open sides, hook & loop closure, 2 mic tabs, radio pocket with trim, inside pocket and built with FR treated solid polyester fabric. Meets ANSI 207-2006, Class 2 and is available in hi-vis yellow with 2" silver heat transferred trim sewn on hi-vis orange gro-grain. Premium Design Features: Radio Pocket with Trim 2 Mic Tabs Inside Pocket 5 point breakaway style Open sides Hook & Loop closure
Catalog No. 50-284-0454