Learn More
Lakeland Industries High Visibility
Supplier:  Lakeland Industries AJPU10LYMD
Encompass_Preferred
Lakeland Arc X FR Rated Polyurethane Rainwear Jackets are comfortable and durable rainwear that offers protection against flash fire and the harsh elements. Available in sizes SM-5X, this lighter weight jacket is ideal for working in cold, rainy weather when flash fires are of concern.
Catalog No. 50-284-0871