Learn More
Lakeland Industries High Visibility
Supplier:  Lakeland Industries VAMOSC2GBVLR
Encompass_Preferred
Lakeland 5-Point Breakaway Public Safety Vests are designed with FR treated mesh fabric, and reflective material. VAMOSCGBV meets ANSI 207-2006 Class 2. Available in Regular and Oversized, these vests are ideal for worker environments where visibility is important, and when flash fire is a concern.
Catalog No. 50-284-1019