Learn More
Lakeland Industries High Visibility
Supplier: Lakeland Industries VAMOSP2GBLR
4-Point Breakaway Public Safety Premium Vest is designed with open sides, zipper closure, 2 mic tabs, radio pocket with trim, inside pocket and built with FR treated mesh polyester fabric. Meets ANSI 207-2006, Class 2 and is available in hi-vis yellow with 2" silver heat transferred trim sewn on hi-vis orange gro-grain.