Learn More
Description
- Combines absorbency, durability and safety signage into one versatile product
- Designed as an all-purpose absorbent for industrial applications
- Perfect option for high-traffic walkways and foot paths
Specifications
Specifications
| Absorbancy | 11.7 gl/gs |
| Color | Black on yellow |
| Capacity (English) | 21.2 gal. |
| Material | Polypropylene |
| Type | Chemical Sorbent Pad |
| For Use With (Application) | High traffic areas where the risk for slips, trips and falls is greatest |
| Packaging Quantity | 100 Pads/Pk. |
| Dimensions (L x W) | 19 x 15 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.